Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FN1 Mouse anti-Human, Unconjugated, Clone: HFN7.1, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB21192
Description
FN1 Monoclonal antibody specifically detects FN1 in Human samples. It is validated for IHC-P, IF, Flow Cyt.Specifications
FN1 | |
Monoclonal | |
Unconjugated | |
FN1 | |
Mouse | |
100 μg | |
2335 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry | |
HFN7.1 | |
fibronectin 1 | |
CIG, DKFZp686F10164, DKFZp686H0342, DKFZp686I1370, DKFZp686O13149, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF | |
Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction