Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FN3K Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566239 Supplier Invitrogen™ Supplier No. PA566239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLKEEENTVGR RGEGAEPQYV DKFGFHTVTC CGFIPQVNEW QDDWPTFFAR HRLQAQLDLI EKDYADREAR E Highest antigen sequence identity to the following orthologs - mouse 82%, rat 81%.

FN3K catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines followed by Amadori rearrangement. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FN3K
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fn3k
Gene Accession No. Q9H479
Gene Alias 2310074G21Rik; FN3K; Fnsk; fructosamine 3 kinase; fructosamine 3-kinase; fructosamine-3-kinase; Protein-psicosamine 3-kinase FN3K; Protein-ribulosamine 3-kinase FN3K
Gene Symbols Fn3k
Host Species Rabbit
Immunogen Recombinant Human FN3K. Recombinant protein control fragment (Product #RP-106920).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.