Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FN3KRP Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144932 Supplier Invitrogen™ Supplier No. PA5144932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRVFVKVNPK AEARRMFEGE MASLTAILKT NTVKVPKPIK VLDA Antigen sequence identity: Mouse 93%; Rat 89%.

Fructosamine-3-kinase related protein (FN3KRP) is located on chromosome 17q25.3, approximately 8 kb upstream of the FN3K gene. It shares 65% amino acid identity with FN3K and possesses an identical genomic organization, implying functional similarities; FN3KRP is suggested to play a role in enzymatic deglycation. Unlike FN3K, FN3KRP is more ancient, present in diverse organisms such as cyanobacteria, echinoderms, plants, and cold-blooded vertebrates. Studies have discovered a significant association between rs1046896 in FN3KRP and longevity, where the longevity allele C is linked to increased FN3KRP expression in whole blood. FN3KRP is hypothesized to phosphorylate glucitolamines, products of non-enzymatic glycation, indicating its involvement in crucial metabolic processes. The regulation of both FN3K and FN3KRP is notable for TATA-less and CAAT-less promoters, featuring homologous CpG islands and Sp1 binding sites. Meanwhile, both genes are widely expressed across human tissues, but FN3K demonstrates significantly higher levels in tissues prone to glycation-related complications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FN3KRP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FN3KRP
Gene Accession No. Q9HA64
Gene Alias FN3KL; FN3K-related protein; Fn3krp; FN3K-RP; fructosamine 3 kinase related protein; fructosamine 3-kinase-related protein; fructosamine-3-kinase-related protein; Ketosamine-3-kinase; ortholog of human fructosamine-3-kinase-related protein FN3KRP; Protein-psicosamine 3-kinase FN3KRP; RGD1304570; testis secretory sperm-binding protein Li 211a
Gene Symbols FN3KRP
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 36-79. Recombinant protein control fragment (Product #RP-109816).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.