Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FN3KRP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730868 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0868 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0868 Supplier Proteintech Group Inc Supplier No. 163121AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Product No. 16312-1-AP is a rabbit polyclonal antibody raised against the full-length of human FN3KRP. The MW of this protein is 34 kDa, and this antibody specifically recognizes the 34 kDa protein.

FN3KRP and FN3K.

Specifications

Antigen FN3KRP
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FN3KRP
Gene Accession No. Q8K274, Q9HA64
Gene Alias FN3K related protein, FN3K RP, FN3KL, FN3KRP, Ketosamine 3 kinase
Gene Symbols FN3KRP
Host Species Rabbit
Immunogen FN3KRP Fusion Protein Ag9347
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238024, 303755, 79672
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.