Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNBP1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147260APC
Description
FNBP1 Polyclonal antibody specifically detects FNBP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, MicroarraySpecifications
FNBP1 | |
Polyclonal | |
Western Blot, Immunoprecipitation, Microarray | |
FBP17KIAA0554Formin-binding protein 17, formin binding protein 1, formin-binding protein 1, hFBP17, MGC126804 | |
The immunogen for this product maps to a region between residue 350 and 400 of human Formin-Binding Protein 17 using the numbering given in entry NP_055848.1 (GeneID 23048). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Microarray | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23048 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction