Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582688 Supplier Invitrogen™ Supplier No. PA582688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KARQQAQIRH QMAEDSKADY SSILQKFNHE QHEYYHTHIP NIFQKIQEME ERRIVRMGES MKTYAEVDRQ VIPIIGKCLD GIVKAAESID QKNDSQLVIE AYKSGFE Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 93%.

The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNBP1
Gene Accession No. Q96RU3
Gene Alias 1110057E06Rik; 2210010H06Rik; FBP1; FBP17; Fnbp1; formin binding protein 1; formin binding protein 17; formin-binding protein 1; Formin-binding protein 17; hFBP17; Kiaa0554; MGC126804; Rapostlin
Gene Symbols FNBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNBP1. Recombinant protein control fragment (Product #RP-92576).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23048
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.