Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FNBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110852
Description
Immunogen sequence: DYTQPMKRTV SDNSLSNSRG EGKPDLKFGG KSKGKLWPFI KKNKLSLKLG ATPEDFSNLP PEQ.
The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.
Specifications
FNBP1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96RU3 | |
FNBP1 | |
Recombinant Protein corresonding to Human FNBP1. Recombinant protein control fragment (Product #RP-108188). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
FNBP1 | |
1110057E06Rik; 2210010H06Rik; FBP1; FBP17; Fnbp1; formin binding protein 1; formin binding protein 17; formin-binding protein 1; Formin-binding protein 17; hFBP17; Kiaa0554; MGC126804; Rapostlin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23048 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction