Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNBP1L/TOCA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981713
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1713 Supplier Bioss Supplier No. BS11841R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination.

Specifications

Antigen FNBP1L/TOCA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FNBP1L
Gene Accession No. Q5T0N5
Gene Alias 2610318I01Rik; AW548221; C1orf39; Cdc42 effector; FNBP1L; formin binding protein 1 like; formin binding protein 1-like; Formin-binding protein 1-like; RGD1305386; Toca1; Toca-1; transducer of Cdc42-dependent actin assembly 1; Transducer of Cdc42-dependent actin assembly protein 1
Gene Symbols FNBP1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FNBP1L/TOCA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54874
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.