Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNBP4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074644
Description
FNBP4 Polyclonal specifically detects FNBP4 in Human samples. It is validated for Immunoprecipitation.Specifications
FNBP4 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DKFZp686M14102, FBP30, FLJ41904, formin binding protein 4, Formin-binding protein 30, formin-binding protein 4, KIAA1014DKFZp779I1064 | |
Rabbit | |
Affinity Purified | |
RUO | |
23360 | |
Human | |
IgG |
Immunoprecipitation | |
1.0 mg/mL | |
Immunoprecipitation 2-5ug/mg Lysate | |
EAW67887.1 | |
FNBP4 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human formin binding protein 4 (formin-binding protein 30) using the numbering given in entry EAW67887.1 (GeneID 23360). | |
100 μL | |
Primary | |
Tested to reaBased on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset, African elephant and Thirteen-lined ground squirrel. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction