Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNBP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568192 Supplier Invitrogen™ Supplier No. PA568192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

FNBP4 encodes a protein containing two tryptophan-rich WW domains that binds the proline-rich formin homology 1 domains of formin family proteins, suggesting a role in the regulation of cytoskeletal dynamics during cell division and migration. It also binds intersectin family proteins suggesting a role in the maintenance of membrane curvature at sites of nascent vesicle formation. Naturally occurring mutations in this gene are associated with Waardenburg anophthalmia syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNBP4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FNBP4
Gene Accession No. Q6ZQ03, Q8N3X1
Gene Alias FBP30; Fnbp30; Fnbp4; formin binding protein 30; formin binding protein 4; formin-binding protein 30; Formin-binding protein 4; Kiaa1014; mKIAA1014
Gene Symbols FNBP4
Host Species Rabbit
Immunogen Synthesized peptide derived from human FNBP4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23360, 55935
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.