Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556602 Supplier Invitrogen™ Supplier No. PA556602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSHPKLSSGI HGDEEDEKPL PATVVNDHVP SSSRQPISRG WEDLRRSPQR GASLHRKEPI PENPKSTGAD THPQGKYSSL A Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 57%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FNDC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNDC1
Gene Accession No. Q4ZHG4
Gene Alias 1110027O12Rik; 1110051A18Rik; Activation-associated cDNA protein; activator of G-protein signaling 8; AGS8; bA243O10.1; dJ322A24.1; expressed in synovial lining protein; fibronectin type III domain containing 1; fibronectin type III domain containing 2; fibronectin type III domain-containing protein 1; FNDC1; FNDC2; KIAA1866; MEL4B3; mKIAA1866
Gene Symbols FNDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNDC1. Recombinant protein control fragment (Product #RP-94940).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84624
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.