Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNDC3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP188810
Description
FNDC3A Polyclonal specifically detects FNDC3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FNDC3A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
bA203I16.1, bA203I16.5, fibronectin type III domain containing 3, fibronectin type III domain containing 3A, fibronectin type-III domain-containing protein 3A, FNDC3, HUGO, Human gene expressed in odontoblasts, KIAA0970FLJ31509 | |
Rabbit | |
Affinity Purified | |
RUO | |
22862 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FNDC3A | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VNWEVPLSNGTDVTEYRLEWGGVEGSMQICYCGPGLSYEIKGLSPATTYYCRVQALSVVGAGPFSEVVACVTPPSVPGIVTCLQEISDDEIENPHYSPSTCLAISWEKPCDHGSEILAYSIDFGDKQSLTVGKV | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction