Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FNDC3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552990
Description
Immunogen sequence: VNWEVPLSNG TDVTEYRLEW GGVEGSMQIC YCGPGLSYEI KGLSPATTYY CRVQALSVVG AGPFSEVVAC VTPPSVPGIV TCLQEISDDE IENPHYSPST CLAISWEKPC DHGSEILAYS IDFGDKQSLT VGKV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
FNDC3A gene ontology annotations related to this gene include cell-cell adhesion; fertilization; Sertoli cell development; spermatid development.
Specifications
FNDC3A | |
Polyclonal | |
Unconjugated | |
FNDC3A | |
1700094E19Rik; bA203I16.1; bA203I16.5; D14Ertd453e; F730017H24Rik; fibronectin domain-containing protein 3a; fibronectin type III domain containing 3a; fibronectin type-III domain-containing protein 3A; FNDC3; fndc3a; HUGO; Human gene expressed in odontoblasts; KIAA0970; sys; wu:fk65b04; zmp:0000001126 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2H6 | |
FNDC3A | |
Recombinant protein corresponding to Human FNDC3A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction