Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNDC3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552630 Supplier Invitrogen™ Supplier No. PA552630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAGDMTTQYM PQYQSSQVYG DVDAHSTHGR SNFRDERSSK TYERLQKKLK DRQGTQKDKM SSPPSSPQKC PSPINEHNGL IKGQIAGGIN TGSAKIKSGK GKGGTQVDTE IEEKDEETKA FEALLSNIVK PVA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 77%.

FNDC3A gene ontology annotations related to this gene include cell-cell adhesion; fertilization; Sertoli cell development; spermatid development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNDC3A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNDC3A
Gene Accession No. Q9Y2H6
Gene Alias 1700094E19Rik; bA203I16.1; bA203I16.5; D14Ertd453e; F730017H24Rik; fibronectin domain-containing protein 3a; fibronectin type III domain containing 3a; fibronectin type-III domain-containing protein 3A; FNDC3; fndc3a; HUGO; Human gene expressed in odontoblasts; KIAA0970; sys; wu:fk65b04; zmp:0000001126
Gene Symbols FNDC3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNDC3A. Recombinant protein control fragment (Product #RP-90047).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.