Learn More
FNDC3A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305158AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
FNDC3A gene ontology annotations related to this gene include cell-cell adhesion; fertilization; Sertoli cell development; spermatid development.Specifications
FNDC3A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y2H6 | |
FNDC3A | |
Between 700 and 750 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FNDC3A | |
bA203I16.1, bA203I16.5, fibronectin type III domain containing 3, FNDC3, HUGO, human gene expressed in odontoblasts, KIAA0970 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22862 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.