Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC3A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982692
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2692 Supplier Bioss Supplier No. BS13191R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FNDC3A gene ontology annotations related to this gene include cell-cell adhesion; fertilization; Sertoli cell development; spermatid development.

Specifications

Antigen FNDC3A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FNDC3A
Gene Accession No. Q9Y2H6
Gene Alias 1700094E19Rik; bA203I16.1; bA203I16.5; D14Ertd453e; F730017H24Rik; fibronectin domain-containing protein 3a; fibronectin type III domain containing 3a; Fibronectin type-III domain-containing protein 3A; FNDC3; Fndc3a; HUGO; Human gene expressed in odontoblasts; Kiaa0970; sys; wu:fk65b04
Gene Symbols FNDC3A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FNDC3A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22862
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.