Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNDC3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543419 Supplier Invitrogen™ Supplier No. PA543419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RARSSPKSND SDLQEYELEV KRVQDILSGI EKPQVSNIQA RAVVLSWAPP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

FNDC3B may be a positive regulator of adipogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNDC3B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FNDC3B
Gene Accession No. Q53EP0
Gene Alias 1600019O04Rik; AW550168; factor for adipocyte differentiation 104; Fad104; fibronectin domain-containing protein 3b; fibronectin type III domain containing 3B; fibronectin type III domain-containing protein 3B; Fndc3b; HCV NS5A-binding protein 37; Kiaa4164; mKIAA4164; Ns5abp37; PRO4979; RGD1311673; UNQ2421/PRO4979/PRO34274; YVTM2421
Gene Symbols FNDC3B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FNDC3B (aa 250-299).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64778
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.