Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNDC3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552474 Supplier Invitrogen™ Supplier No. PA552474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIPPIHVPPG YISQVIEDST GVRRVVVTPQ SPECYPPSYP SAMSPTHHLP PYLTHHPHFI HNSHTAYYPP VTGPGDMPPQ FFPQHHLPHT IYGEQEIIPF YGMSTYITRE DQYSK Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.

FNDC3B may be a positive regulator of adipogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNDC3B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNDC3B
Gene Accession No. Q53EP0, Q6NWW9
Gene Alias 1600019O04Rik; AW550168; factor for adipocyte differentiation 104; Fad104; fibronectin domain-containing protein 3b; fibronectin type III domain containing 3B; fibronectin type III domain-containing protein 3B; Fndc3b; HCV NS5A-binding protein 37; Kiaa4164; mKIAA4164; Ns5abp37; PRO4979; RGD1311673; UNQ2421/PRO4979/PRO34274; YVTM2421
Gene Symbols FNDC3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNDC3B. Recombinant protein control fragment (Product #RP-89832).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294925, 64778, 72007
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.