Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNDC4 Antibody (OTI1B7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272151FR
Description
FNDC4 Monoclonal specifically detects FNDC4 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
FNDC4 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
FNDC4 | |
Full length recombinant protein of human FNDC4 (NP_073734) produced in HEK293T cells. | |
0.1 mL | |
64838.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI1B7 | |
Immunohistochemistry | |
fibronectin type III domain containing 4, fibronectin type III domain-containing protein 4, Fibronectin type III repeat-containing protein 1, FRCP1FLJ22362 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction