Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNDC4 Antibody (OTI1B7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245691
Description
Ensure accurate, reproducible results in Immunohistochemistry
FNDC4 Monoclonal specifically detects FNDC4 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FNDC4 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1:150 | |
fibronectin type III domain containing 4, fibronectin type III domain-containing protein 4, Fibronectin type III repeat-containing protein 1, FRCP1FLJ22362 | |
Mouse | |
25 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry | |
OTI1B7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
FNDC4 | |
Full length human recombinant protein of human FNDC4(NP_073734) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
64838 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction