Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNDC4 Monoclonal Antibody (OTI1B7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505463S


Description
FNDC4 Monoclonal antibody specifically detects FNDC4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FNDC4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H6D8 | |
Mouse | |
Affinity chromatography | |
RUO | |
64838 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin) | |
OTI1B7 | |
Unconjugated | |
FNDC4 | |
2810430J06Rik; 6330410H20Rik; AB030187; AI838506; AW487863; fibronectin type III domain containing 4; fibronectin type III domain-containing protein 4; fibronectin type III repeat containing protein 1; fibronectin type III repeat-containing protein 1; Fndc4; Fnmp1; FRCP1; UNQ6389/PRO21134 | |
Full length human recombinant protein of human FNDC4 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction