Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC4 Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
SDP

Catalog No. 502539692
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9692 30 μL
50-167-5932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9692 Supplier OriGene Supplier No. TA505459S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FNDC4 Monoclonal antibody specifically detects FNDC4 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen FNDC4
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI3E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GAPDHS
Gene Accession No. O14556
Gene Alias 2810430J06Rik; 6330410H20Rik; AB030187; AI838506; AW487863; fibronectin type III domain containing 4; fibronectin type III domain-containing protein 4; fibronectin type III repeat containing protein 1; fibronectin type III repeat-containing protein 1; Fndc4; Fnmp1; FRCP1; UNQ6389/PRO21134
Host Species Mouse
Immunogen Full length human recombinant protein of human GAPDHS produced in HEK293T
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26330
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.