Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC4 Mouse anti-Human, Clone: OTI3B11, lyophilized, TrueMAB™
SDP

Catalog No. 501663655
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-3655 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-3655 Supplier OriGene Supplier No. CF505462
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Acts as an anti-inflammatory factor in the intestine and colon. Binds to and acts on macrophages to downregulate pro-inflammatory gene expression. Affects key macrophage functions, including phagocytosis, by downregulating many key pathways for macrophage activation, partly via by STAT3 activation and signaling. May be required to dampen the immunological response in colitis.

Specifications

Antigen FNDC4
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI3B11
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene FNDC4
Gene Accession No. Q9H6D8
Gene Alias FRCP1
Gene Symbols FNDC4
Host Species Mouse
Immunogen Full length human recombit protein of human FNDC4 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64838
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.