Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNDC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564124 Supplier Invitrogen™ Supplier No. PA564124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPSPVNVTVT HLRANSATVS WDVPEGNIVI GYSISQQRQN GPGQRVIREV NTTT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 63%.

Acts as an anti-inflammatory factor in the intestine and colon. Binds to and acts on macrophages to downregulate pro-inflammatory gene expression. Affects key macrophage functions, including phagocytosis, by downregulating many key pathways for macrophage activation, partly via by STAT3 activation and signaling. May be required to dampen the immunological response in colitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNDC4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNDC4
Gene Accession No. Q9H6D8
Gene Alias 2810430J06Rik; 6330410H20Rik; AB030187; AI838506; AW487863; fibronectin type III domain containing 4; fibronectin type III domain-containing protein 4; fibronectin type III repeat containing protein 1; fibronectin type III repeat-containing protein 1; Fndc4; Fnmp1; FRCP1; UNQ6389/PRO21134
Gene Symbols FNDC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNDC4. Recombinant protein control fragment (Product #RP-103724).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64838
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.