Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982693
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2693 Supplier Bioss Supplier No. BS13192R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acts as an anti-inflammatory factor in the intestine and colon. Binds to and acts on macrophages to downregulate pro-inflammatory gene expression. Affects key macrophage functions, including phagocytosis, by downregulating many key pathways for macrophage activation, partly via by STAT3 activation and signaling. May be required to dampen the immunological response in colitis.

Specifications

Antigen FNDC4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FNDC4
Gene Accession No. Q9H6D8
Gene Alias 2810430J06Rik; 6330410H20Rik; AB030187; AI838506; AW487863; fibronectin type III domain containing 4; Fibronectin type III domain-containing protein 4; fibronectin type III repeat containing protein 1; Fibronectin type III repeat-containing protein 1; Fndc4; Fnmp1; Frcp1; UNQ6389/PRO21134
Gene Symbols FNDC4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FNDC4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64838
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.