Learn More
FNDC5 Monoclonal Antibody (1736CT532.85.53.92)

Description
FNDC5 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | FNDC5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1736CT532.85.53.92 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FNDC5 |
| Gene Accession No. | Q8NAU1 |
| Gene Alias | 1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.