Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNDC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562368 Supplier Invitrogen™ Supplier No. PA562368
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDTEYIVHVQ AISIQGQSPA SEPVLFKTPR EAEKMASKNK DEVTMKEMGR NQQLR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNDC5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNDC5
Gene Accession No. Q8NAU1
Gene Alias 1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp
Gene Symbols FNDC5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNDC5. Recombinant protein control fragment (Product #RP-101405).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252995
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.