Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNDC8 Antibody (OTI1C10), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271966AF488
Description
FNDC8 Monoclonal specifically detects FNDC8 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| FNDC8 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FNDC8 | |
| Full length recombinant protein of human FNDC8 (NP_060029) produced in HEK293T cells. | |
| 0.1 mL | |
| 54752.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1C10 | |
| Western Blot, Immunohistochemistry | |
| DKFZp434H2215, fibronectin type III domain containing 8, fibronectin type III domain-containing protein 8 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction