Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNDC8 Antibody (OTI1C10), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271966AF750
Description
FNDC8 Monoclonal specifically detects FNDC8 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FNDC8 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
FNDC8 | |
Full length recombinant protein of human FNDC8 (NP_060029) produced in HEK293T cells. | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1C10 | |
Western Blot, Immunohistochemistry | |
DKFZp434H2215, fibronectin type III domain containing 8, fibronectin type III domain-containing protein 8 | |
Mouse | |
0.1 mL | |
54752 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction