Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNDC8 Antibody (OTI1C10), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271966FR
Description
FNDC8 Monoclonal specifically detects FNDC8 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FNDC8 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
FNDC8 | |
Full length recombinant protein of human FNDC8 (NP_060029) produced in HEK293T cells. | |
0.1 mL | |
54752.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1C10 | |
Western Blot, Immunohistochemistry | |
DKFZp434H2215, fibronectin type III domain containing 8, fibronectin type III domain-containing protein 8 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction