Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FNDC9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569303
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 92%; Human: 93%; Mouse: 86%; Rabbit: 93%; Rat: 93%.
FNDC9 is a protein coding gene.
Specifications
FNDC9 | |
Polyclonal | |
Unconjugated | |
FNDC9 | |
C030019I05Rik; C5orf40; fibronectin type III domain containing 9; fibronectin type III domain-containing protein 9; fibronectin type-III domain-containing protein C5orf40; fibronectin type-III domain-containing protein C5orf40 homolog; Fndc9; rCG33426-like | |
Rabbit | |
Affinity chromatography | |
RUO | |
320116 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BJN4 | |
FNDC9 | |
synthetic peptide directed towards the middle region of human FNDC9 (aa 11-60). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction