Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FNIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564359
Description
Immunogen sequence: HHTKPLKEER GAIDQHQETK QTTKDQSGES DTQNMVSEEP CELPCWNHSD PESMSLFDEY FNDDSIETRT IDDVPF Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
FNIP1 encodes a protein that binds to the tumor suppressor protein folliculin and to AMP-activated protein kinase (AMPK). The encoded protein participates in the regulation of cellular metabolism and nutrient sensing by modulating the AMPK and target of rapamycin signaling pathways. This gene has a closely related paralog that encodes a protein with similar binding activities. Both related proteins also associate with the molecular chaperone heat shock protein-90 (Hsp90) and negatively regulate its ATPase activity and facilitate its association with folliculin. Diseases associated with FNIP1 include Birt-Hogg-Dube Syndrome and Pleural Disease.
Specifications
FNIP1 | |
Polyclonal | |
Unconjugated | |
FNIP1 | |
A730024A03Rik; AI838773; AW557298; FNIP1; folliculin interacting protein 1; folliculin-interacting protein 1; Kiaa1961 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
96459 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TF40 | |
FNIP1 | |
Recombinant protein corresponding to Human FNIP1. Recombinant protein control fragment (Product #RP-104226). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction