Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571423 Supplier Invitrogen™ Supplier No. PA571423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FNIP1 Polyclonal Antibody for Western Blot

FNIP1 encodes a protein that binds to the tumor suppressor protein folliculin and to AMP-activated protein kinase (AMPK). The encoded protein participates in the regulation of cellular metabolism and nutrient sensing by modulating the AMPK and target of rapamycin signaling pathways. This gene has a closely related paralog that encodes a protein with similar binding activities. Both related proteins also associate with the molecular chaperone heat shock protein-90 (Hsp90) and negatively regulate its ATPase activity and facilitate its association with folliculin. Diseases associated with FNIP1 include Birt-Hogg-Dube Syndrome and Pleural Disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNIP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FNIP1
Gene Accession No. Q8TF40
Gene Alias A730024A03Rik; AI838773; AW557298; FNIP1; folliculin interacting protein 1; folliculin-interacting protein 1; Kiaa1961
Gene Symbols FNIP1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FNIP1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 96459
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.