Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNIP2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281941MFV610
Description
FNIP2 Polyclonal antibody specifically detects FNIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| FNIP2 | |
| Polyclonal | |
| Western Blot, ELISA | |
| FNIP1-like protein, FNIPL, folliculin interacting protein 2 | |
| FNIP2 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human FNIP2. The immunogen is located within amino acids 100 - 150 of FNIP2. Amino Acid Squence: HHAKEQLPKYQYTRPASD | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 57600 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction