Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520691
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520691 Supplier Invitrogen™ Supplier No. PA520691
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse skeletal muscle tissue lysate. PA5-20691 can be used with blocking peptide PEP-0806.

FNIP2 is the second protein found to interact with folliculin, the product of the Birt-Hogg-Dube (BHD) gene. Folliculin is thought to act as a tumor suppressor as mutations or loss of heterozygosity in this gene are associated with BHD syndrome-related renal tumors. Folliculin and FNIP1, a protein that shares 49% identity to FNIP2, bind to AMPK, an important energy sensor in cells that negatively regulates the mammalian target of rapamycin (mTOR), a protein that is thought to be the master switch for cell growth and proliferation. FNIP1 and FNIP2 are able to form homo- and heteromeric multimers, suggesting these proteins may have a functional relationship.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNIP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FNIP2
Gene Accession No. Q80TD3, Q9P278
Gene Alias D630023B12Rik; FNIP1-like protein; Fnip2; Fnipl; folliculin interacting protein 2; folliculin-interacting protein 2; KIAA1450; MAPO1; mKIAA1450; O6-methylguanine-induced apoptosis 1 protein; RGD1562174
Gene Symbols FNIP2
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human FNIP2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310538, 329679, 57600
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.