Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNTA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | FNTA |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056500T
![]() |
Novus Biologicals
NB10056500SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056500
![]() |
Novus Biologicals
NB10056500 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
FNTA Polyclonal specifically detects FNTA in Human, Rat, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FNTA | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Rat, Canine, Chicken, Primate | |
P49354 | |
2339 | |
A portion of amino acids 150-250 of human farnesyltransferase was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
CAAX farnesyltransferase subunit alpha, EC 2.5.1.58, EC 2.5.1.59, farnesyl-protein transferase alpha-subunit, farnesyltransferase, CAAX box, alpha, FPTA, FTase-alpha, GGTase-I-alpha, MGC99680, PGGT1A, protein farnesyltransferase/geranylgeranyltransferase type-1 subunit alpha, protein prenyltransferase alpha subunit repeat containing 2, PTAR2, Ras proteins prenyltransferase subunit alpha, type I protein geranyl-geranyltransferase alpha subunit, Type I protein geranyl-geranyltransferase subunit alpha | |
FNTA | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, _b, _c) and chimp (isoforms 1, 2, 3) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title