Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNTA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553858 Supplier Invitrogen™ Supplier No. PA553858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDSPSYVLYR HFRRVLLKSL QKDLHEEMNY ITAIIEEQPK NYQVWHHRRV LVEWLRDPSQ ELEFIADI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

FNTA (Farnesyltransferase, CAAX box; alphaCAAX farnesyltransferase alpha subunit, FPTA, FTase-alpha, GGTase-I-alpha, Protein farnesyl transferase/ geranyl geranyl transferase type I alpha subunit(PGGT1A), Ras proteins prenyl transferase alpha, Type I protein geranyl-geranyl transferase alpha subunit) is located locus 8p11 (human). Prenylation is a common intracellular posttranslational protein modification catalyzed by prenyltransferases. The best characterized among these prenyltransferases is the CAAX farnesyl transferase (2. 5. 1. 21), which is alpha-beta heterodimeric enzyme. It catalyzes the attachment of a farnesyl group from farnesyl phosphate to cysteine residues at the fourth position from the C terminus of proteins that end in the CAAX box. FNTA or the alpha subunit of farnesyl transferase is shared by the geranyl-geranyl protein transferases. Apart from transfer of farnesyl and geranyl groups to proteins, FNTA has been proposed to regulate intracellular signaling pathways involving p21ras. Reports have shown that FNTA interacts with TGF-beta and activin-type I receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNTA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNTA
Gene Accession No. P49354
Gene Alias CAAX farnesyltransferase subunit alpha; farnesyl-protein transferase alpha-subunit; farnesyltransferase, CAAX box, alpha; Farnesyltransferase, subunit alpha; Fnta; FPTA; FTA; FTase-alpha; GGTase-I-alpha; PFAS; PGGT1A; Protein farnesyltransferase/geranylgeranyltransferase type-1 subunit alpha; protein prenyltransferase alpha subunit repeat containing 2; PTAR2; Ras proteins prenyltransferase subunit alpha; type I protein geranyl-geranyltransferase alpha subunit; type I protein geranyl-geranyltransferase subunit alpha
Gene Symbols FNTA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNTA. Recombinant protein control fragment (Product #RP-101897).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2339
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.