Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FNTB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214960
Description
FNTB Polyclonal specifically detects FNTB in Human samples. It is validated for Western Blot.Specifications
FNTB | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
CAAX farnesyltransferase subunit beta, EC 2.5.1.58, farnesyltransferase, CAAX box, beta, FPTB, FTase-beta, MGC31935, protein farnesyltransferase subunit beta, Ras proteins prenyltransferase subunit beta | |
Rabbit | |
49 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
0.29mg/mL | |
Western Blot 1:500-1:3000 | |
P49356 | |
FNTB | |
Recombinant protein encompassing a sequence within the center region of human FNTB. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
2342 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction