Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNTB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597757
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597757 Supplier Invitrogen™ Supplier No. PA597757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FNTB Polyclonal Antibody for ELISA, ICC/IF, IHC (P), IP, Western Blot

Eukaryotic cells contain 3 different types of prenyltransferases that attach either a farnesyl group (15 carbons) or a geranylgeranyl group (20 carbons) in thioether linkage to C-terminal cysteine residues in a variety of proteins. These posttranslational modifications provide a mechanism for membrane localization of proteins that lack a transmembrane domain. CAAX farnesyltransferase (FTase) attaches a farnesyl group from farnesyl pyrophosphate to cysteine residues at the fourth position from the C terminus of proteins that end in the CAAX box, where C is cysteine, A is usually but not always an aliphatic amino acid, and X is typically methionine or serine. This enzyme has the ability to farnesylate peptides as short as 4 residues in length that conform to the CAAX consensus sequence. The gene for the beta subunit of CAAX farnesyltransferase (FNTB) has been pinpointed to 14q23-q24 by Southern blot hybridization and PCR analyses of panels of human/Chinese hamster somatic cell hybrid lines and by fluorescence chromosomal in situ hybridization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNTB
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FNTB
Gene Accession No. P49356
Gene Alias 2010013E13Rik; AA409500; CAAX farnesyltransferase subunit beta; EC 2.5.1.58; farnesyl-protein transferase beta-subunit; farnesyltransferase beta subunit; farnesyltransferase, CAAX box, beta; FNTB; FPTB; ft beta; FTase beta; FTase-beta; LOW QUALITY PROTEIN: protein farnesyltransferase subunit beta; protein farnesyltransferase subunit beta; Ras proteins prenyltransferase subunit beta
Gene Symbols FNTB
Host Species Rabbit
Immunogen Recombinant Human Protein farnesyltransferase subunit beta protein (1-300AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2342
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.