Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FNTB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564061 Supplier Invitrogen™ Supplier No. PA564061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASPSSFTYY CPPSSSPVWS EPLYSLRPEH ARERLQDDSV ETVTSIEQAK VEEK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.

Eukaryotic cells contain 3 different types of prenyltransferases that attach either a farnesyl group (15 carbons) or a geranylgeranyl group (20 carbons) in thioether linkage to C-terminal cysteine residues in a variety of proteins. These posttranslational modifications provide a mechanism for membrane localization of proteins that lack a transmembrane domain. CAAX farnesyltransferase (FTase) attaches a farnesyl group from farnesyl pyrophosphate to cysteine residues at the fourth position from the C terminus of proteins that end in the CAAX box, where C is cysteine, A is usually but not always an aliphatic amino acid, and X is typically methionine or serine. This enzyme has the ability to farnesylate peptides as short as 4 residues in length that conform to the CAAX consensus sequence. The gene for the beta subunit of CAAX farnesyltransferase (FNTB) has been pinpointed to 14q23-q24 by Southern blot hybridization and PCR analyses of panels of human/Chinese hamster somatic cell hybrid lines and by fluorescence chromosomal in situ hybridization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNTB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNTB
Gene Accession No. P49356
Gene Alias 2010013E13Rik; AA409500; CAAX farnesyltransferase subunit beta; EC 2.5.1.58; farnesyl-protein transferase beta-subunit; farnesyltransferase beta subunit; farnesyltransferase, CAAX box, beta; FNTB; FPTB; ft beta; FTase beta; FTase-beta; LOW QUALITY PROTEIN: protein farnesyltransferase subunit beta; protein farnesyltransferase subunit beta; Ras proteins prenyltransferase subunit beta
Gene Symbols FNTB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FNTB. Recombinant protein control fragment (Product #RP-103467).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.