Learn More
StatLab™ Foam Biopsy Sponge
SDP
Catalog No. p-7227792
Click to view available options
Color:
Black
Blue
White
This item is not returnable. View return policy
This item is not returnable. View return policy
Thin foam pad that secures small specimens during processing.
  • Aid with preventing biopsies from being lost during processing
Specifications
Color Blue
Length (English) 1 in.
Width (English) 11/4 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.