Learn More
Foamtec ScrubWRIGHT™ Chamber Cleaning Tool
Shop All Foamtec International Products
Click to view available options
Grit:
1350
280
360
800
Not Applicable
Product Type:
Scrub Belt
Scrub Pen
Description
- Removes deposition from areas critical to process
- Prevents unwanted flaking
- High impact plastic w/spring-tensioned attachment mechanism
- Interchangeable belt-shaped
- Spring adjustable for easy mounting and rotation
- Minimizes tool wear
- For polishing stains, scratches and process-induced residue from scratch sensitive surfaces
Specifications
Specifications
Product Type | Scrub Pen |
For Use With (Application) | For scrubbing hard-to-reach areas, tight corners |
Grit | Not Applicable |
Includes | Spring tensioned attachment mechanism |
Material | Plastic |