Learn More
Foamtec UltraMOP™ II Mop Heads
Shop All Foamtec International Products
Click to view available options
Product Type:
Non-laminated Mop Head
Polyester Laminated Mop Head
Refill
Sterility:
Non-sterile
Sterile
Description
Constructed from a fine-pore urethane foam to enhance application of disinfectants to clean room surfaces. Available in laminated versions to allow tear-free cleaning and disinfecting of areas that have rough or sharp surfaces.
- Proprietary polyurethane foam compatible with disinfectants used for maintaining sterile environments
- Laminated bump-proof designs enable tear-free cleaning and disinfecting of areas that have rough or sharp surfaces
- Micro-fiber versions are designed to remove and absorb stubborn contamination such as hardened biofilms and disinfectant residue, perfect for drying floors
- Individually packaged to maintain cleanliness
- Fine, uniform pore size and proprietary, patent-pending, lamination techniques ensure excellent laydown and pickup properties
- Available with lot Certification of Conformance for ease of audit and validation procedures
Specifications
Specifications
| Product Type | Polyester Laminated Mop Head |
| Length (English) | 16 in. |
| Sterility | Sterile |
| Includes | Polyester Lamination |
| For Use With (Application) | Designed for wet mopping procedures in sterile and controlled environments |
| Material | Urethane Foam |