Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOG2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981804
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1804 Supplier Bioss Supplier No. BS11942R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The zinc finger protein encoded by this gene is a widely expressed member of the FOG family of transcription factors. The family members modulate the activity of GATA family proteins, which are important regulators of hematopoiesis and cardiogenesis in mammals. It has been demonstrated that the protein can both activate and down-regulate expression of GATA-target genes, suggesting different modulation in different promoter contexts. A related mRNA suggests an alternatively spliced product but this information is not yet fully supported by the sequence.

Specifications

Antigen FOG2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZFPM2
Gene Accession No. Q8WW38
Gene Alias B330005D23Rik; DIH3; Fog2; FOG-2; Friend of GATA 2; Friend of GATA protein 2; friend of GATA-1 2; Friend of GATA2; friend of GATA-2; hFOG-2; mFOG-2; SRXY9; transcription factor GATA4, modulator of; ZC2HC11B; Zfpm2; Zinc finger protein 89B; Zinc finger protein multitype 2; Zinc finger protein ZFPM2; zinc finger protein, FOG family member 2; zinc finger protein, multitype 2; ZNF89B
Gene Symbols ZFPM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FOG2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23414
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.