Learn More
Folate Binding Protein Polyclonal Antibody, Biotin, Rockland™

Goat Polyclonal Antibody
$178.35 - $626.07
Specifications
| Antigen | Folate Binding Protein |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0394
![]()
|
Rockland Immunochemicals
2001061480100 |
100 μg |
Each for $626.07
|
|
|||||
50-199-0395
![]()
|
Rockland Immunochemicals
200106148S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Folate Binding Protein is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Goat Serum as well as purified and partially purified Folate Binding Protein [Bovine Milk]. Cross reactivity against Folate Binding Protein from other sources is unknown.
The protein encoded by this gene is a member of the folate receptor family. Members of this gene family bind folic acid and its reduced derivatives, and transport 5-methyltetrahydrofolate into cells. This gene product is a secreted protein that either anchors to membranes via a glycosyl-phosphatidylinositol linkage or exists in a soluble form. Mutations in this gene have been associated with neurodegeneration due to cerebral folate transport deficiency. Due to the presence of two promoters, multiple transcription start sites, and alternative splicing, multiple transcript variants encoding the same protein have been found for this gene.Specifications
| Folate Binding Protein | |
| ELISA, Immunoprecipitation, Western Blot | |
| Biotin | |
| Goat | |
| Bovine | |
| P02702 | |
| 539750 | |
| Folate Binding Protein [Bovine Milk] | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FOLR1 |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
| Adult folate-binding protein; BOS_14952; FBP; Fbp1; folate binding protein; folate binding protein 1; Folate receptor 1; folate receptor 1 (adult); Folate receptor alpha; Folate receptor, adult; Folate-binding protein 1; Folbp1; Folbp-1; FOLR; Folr1; FR-alpha; KB cells FBP; LOC609612; MOv18; Ovarian tumor-associated antigen MOv18 | |
| FOLR1 | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.