Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Folate Receptor alpha Monoclonal Antibody (06), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543019
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543019 100 μL
PIMA543380 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543019 Supplier Invitrogen™ Supplier No. MA543019
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human FOLR1

The protein encoded by this gene is a member of the folate receptor family. Members of this gene family bind folic acid and its reduced derivatives, and transport 5-methyltetrahydrofolate into cells. This gene product is a secreted protein that either anchors to membranes via a glycosyl-phosphatidylinositol linkage or exists in a soluble form. Mutations in this gene have been associated with neurodegeneration due to cerebral folate transport deficiency. Due to the presence of two promoters, multiple transcription start sites, and alternative splicing, multiple transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Folate Receptor alpha
Applications ELISA
Classification Monoclonal
Clone 06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene FOLR1
Gene Accession No. P15328
Gene Alias 66/67kDa; AA407891; adult folate-binding protein; eIF3 p66; EIF3D; eIF3d {ECO:0000255; eIF3p66; eIF3-p66; EIF3S7; eIF-3-zeta; eIF3-zeta; eIF-3-zeta {ECO:0000255; Eukaryotic translation initiation factor 3 subunit 7; eukaryotic translation initiation factor 3 subunit 7 {ECO:0000255; eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3 subunit D {ECO:0000255; eukaryotic translation initiation factor 3, subunit 7 (zeta); eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; FBP; Fbp1; folate binding protein; folate binding protein 1; folate receptor 1; folate receptor 1 (adult); folate receptor alpha; Folate receptor, adult; folate-binding protein 1; Folbp1; Folbp-1; FOLR; FOLR1; FR-alpha; HAMAP-Rule:MF_03003}; KB cells FBP; MOv18; Ovarian tumor-associated antigen MOv18; translation initiation factor eIF3 p66; translation initiation factor eIF3 p66 subunit
Gene Symbols FOLR1
Host Species Mouse
Immunogen Recombinant Human FOLR1 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2348
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.