Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Folate Receptor alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5112130

Description
This product can be used in direct ELISA.
Folate Receptor alpha Polyclonal antibody specifically detects Folate Receptor alpha in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Specifications
Folate Receptor alpha | |
Polyclonal | |
Unconjugated | |
FOLR1 | |
Adult folate-binding protein; BOS_14952; FBP; Fbp1; folate binding protein; folate binding protein 1; Folate receptor 1; folate receptor 1 (adult); Folate receptor alpha; Folate receptor, adult; Folate-binding protein 1; Folbp1; Folbp-1; FOLR; Folr1; FR-alpha; KB cells FBP; LOC609612; MOv18; Ovarian tumor-associated antigen MOv18 | |
Recombinant Human FOLR1 Protein | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
P15328 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction