Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Folate Receptors α/β (FRαβ) Rat anti-Human, PE, Clone: 5 (also known as No.5; No. 5; Antibody No.5; Antibody No. 5; 5; 5/FRAB), BD Pharmingen™
Rat Monoclonal Antibody
Supplier: BD Biosciences 567605
This item is not returnable.
View return policy
Description
Folate Receptors α/β (FRαβ) Monoclonal antibody specifically detects Folate Receptors α/β (FRαβ) in Human samples. It is validated for Flow Cytometry
Specifications
Folate Receptors α/β (FRαβ) | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2a κ |
Flow Cytometry | |
5 (also known as No.5; No. 5; Antibody No.5; Antibody No. 5; 5; 5/FRAB) | |
0.02% Sodium Azide | |
FOLR1, Folate receptor 1, FR-alpha, FRalpha; FOLR2, Folate receptor 2, FR-beta, FRbeta; FRαβ, FR-αβ | |
Human Folate Receptor beta Transfected Cell Line | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction