Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Fold-ups Caution Tripping Hazard Sign

Catalog No. 19804780 Shop All AccuformNMC Products
Click to view available options
Color:
Red on Yellow

Identify temporary hazardous conditions or display messages quickly and easily

  • Double-sided signs can be set-up and moved easily to exactly where they are needed
  • Rugged, high-impact plastic construction with handle cut-out
  • Fold flat for easy storage and transport
  • All signs include same text and pictogram on both sides
  • All signs measure 20H x 12 in. W (51 x 30.5cm)

Specifications

Material Plastic
Quantity 1 Ea.
For Use With (Application) Free standing signs allow you to identify temporary hazardous conditions or display important messages, quickly and easily
Height (English) 20 in.
Thickness 0.12 in.
Width (English) 12 in.
Dimensions (L x W) 12 x 20 in.
Displayed Text CAUTION Tripping Hazard
Product Type Fold-up Floor Safety Sign
Color Red on Yellow
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.