Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLH1 Rabbit anti-Human, Unconjugated, Clone: RM327, Abnova™

Catalog No. 89339188 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-188 Supplier Abnova Corporation Supplier No. MAB21971
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

FOLH1 Monoclonal antibody specifically detects FOLH1 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen FOLH1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM327
Conjugate Unconjugated
Description folate hydrolase (prostate-specific membrane antigen) 1
Gene FOLH1
Gene Alias FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human FOLH1
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 2346
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.